missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, USP grade, MP Biomedicals™

Catalog No. p-4589754
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19021610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19021610 Supplier MP Biomedicals Inc Supplier No. 0219021610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.

  • Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.
  • Also inhibits cyclooxygenase, PGH synthase-1 and PGH synthase-2.
  • Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23.

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
PubChem CID 3672
ChEBI CHEBI:5855
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 75 to 77.5°C
Color White
Quantity 10 g
Solubility Information Water Solubility: 21 mg/L (at 25°C)
Vapor Pressure 4.74X10-5 mm Hg at 25°C
Formula Weight 206.3
Grade USP
Physical Form Powder
Chemical Name or Material Ibuprofen
EINECSNumber 239-784-6
Recommended Storage Store at Room Temperature (15 to 30°C)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.