missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen Impurity G Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1391054-15-1 |
| Linear Formula | C26H32O4 |
| Synonym | (1 RS,4 RS)-7-(2-Methylpropyl)-1-[4-(2-methylpropyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic acid |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 408.53 |
| Grade | Certified Reference Material |