missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen , ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013256
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013256 1 g
111013257 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013256 Supplier Merck Emd Millipore Supplier No. I1101G

Ibuprofen is a non-steroidal and anti-inflammatory drug, which finds a variety of therapeutic applications in relieving pain and inflammatory disorders.

Pharmaceutical (small molecule)

Chemical Identifiers

MDL Number MFCD00010393
Synonym alpha-Methyl-4-(isobutyl)phenylacetic acid; (+/-)-2-(4-Isobutylphenyl)propanoic acid

Specifications

CAS 15687-27-1
Packaging Poly Bottle
Linear Formula C13H18O2
Quantity 1 g
Formula Weight 206.28
Percent Purity ≥98% (HPLC)
Physical Form Neat
RTECSNumber MU6640000