missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IBMX

Catalog No. PHZ1124
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHZ1124 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHZ1124 Supplier Invitrogen™ Supplier No. PHZ1124

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IBMX

IBMX is a nonspecific inhibitor of phosphodiesterases that also possesses adenosine agonist activity.

Format/Formulation: Lyophilized

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Gene ID (Entrez) 5595
Purification Method TLC
Shipping Condition Room Temperature
Specific Inhibit or Target Phosphodiesterases (also has adenosine agonist activity)
Sterility Non-sterile
Quantity 1 g
Product Type Inhibitor
Research Category Signal Transduction
Inhibitor Properties Chemical
Molecular Weight (g/mol) 222.2
Inhibitors Phosphodiesterase Inhibitor
Form Solid
Show More Show Less
Do you offer an alternative for the Revvity Health Sciences Inc IBMX Phosphodiesterase inhibitor (Cat. No. 50-211-8316) from Fisher Scientific?

Yes. We offer IBMX (Cat. No. PHZ1124), which can be found linked below:

IBMX (Cat. No. PHZ1124)

For Research Use Only. Not for use in diagnostic procedures.