missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 Western Blot Transfer Device

Catalog No. p-9954003
Change view
Click to view available options
Warranty:
REX Extended Warranty, 2 years
Standard, 1-year warranty included
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Warranty
A56727PKG REX Extended Warranty, 2 years
IB31001 Standard, 1-year warranty included
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A56727PKG Supplier Invitrogen™ Supplier No. A56727

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers.

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

iBlot 3 Western Blot Transfer Device features include:
• Complete protein transfer in as few as three minutes
• High detection sensitivity and even, consistent protein transfer
• Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
• Built-in adjustable cooling to ensure consistent transfer temperature
• Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
• Ability to create custom programs
• A simple, user-friendly interface

How the iBlot dry blotting system works
Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen).

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

Specifications

Content And Storage • iBlot 3 Gel Transfer Device
• Western Blot Roller
• REX Extended Warranty

Store at room temperature.
For Use With (Application) Western Blot
For Use With (Equipment) iBlot 3 Transfer Stacks
Gel Compatibility Mini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 3
Running Time ∼3 to 10 min.
Throughput 2 Midi or 4 Mini Gels
Type Transfer Device
Quantity 1 device with warranty
Warranty REX Extended Warranty, 2 years
Show More Show Less
Are the E-PAGE Gels (example Cat. No. EP09606) compatible with the iBlot3 device (Cat. O. IB31001)?

Due to the thickness of the E-PAGE gels, they are not compatible with the iBlot3 device for transfer. Recommended methods for transfer of E-PAGE gels are semi-dry blotting or semi-wet blotting in the XCell II Blot Module. Methods are described in the E-PAGE Technical Guide.

Do you offer additional years of service for the iBlot 3 Western Transfer Device?

No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.

For Research Use Only. Not for use in diagnostic procedures.