missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 iBind™ Western Starter Kit
GREENER_CHOICE

Catalog No. IB31001SLF1
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IB31001SLF1 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IB31001SLF1 Supplier Invitrogen™ Supplier No. IB31001SLF1
  • ONLINE EXCLUSIVE PRICE
    $8,180.00
    $5,317.00 / Pack of 6 Each

    Save $2,863.00

    35.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The iBlot 3 iBind Western Starter Kit includes the iBlot 3 Western Blot Transfer Device, the iBind Western Device, and all of the consumables and reagents necessary to get started using both devices for fast, efficient transfer and western processing.

The iBlot 3 iBind Western Starter Kit includes the iBlot 3 Western Blot Transfer Device, the iBind Western Device, and all of the consumables and reagents necessary to get started using both devices for fast, efficient transfer and western processing.

Contents of the iBlot 3 iBind Western Starter Kit:

  • iBlot 3 Western Blot Transfer Device
  • iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose)
  • iBind Western Device
  • iBind Cards
  • iBind Solution Kit
  • SuperSignal West Pico Plus Chemiluminescent Substrate

iBlot 3 Western Blot Transfer Device
The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

iBind Western Device
The iBind Western Device is an automated western-processing device that performs every step in the western protocol from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source, shakers, or trays. Each solution is released from the iBind device wells to an iBind Card via SLF in ∼2.5 hours. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction. The iBind western detection procedure minimizes hands-on time and is highly reproducible and compatible with all western protocols.

Specifications

Content And Storage • iBlot 3 Gel Transfer Device
• Western Blot Roller
• iBlot 3 Transfer Stacks, midi, nitrocellulose
• iBind Western Device
• iBind Cards
• iBind Solution Kit
• SuperSignal West Pico Plus Chemiluminescent Substrate

Detection Method Fluorescence, Chemiluminescence, Chromogenic
For Use With (Application) Western Blot
Gel Compatibility Mini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 3, iBind
Type Starter Kit
Quantity 1 kit
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Do you offer additional years of service for the iBlot 3 Western Transfer Device?

No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.