Learn More
Invitrogen™ iBind™ Flex Western Starter Kit, fluorescent detection
The iBind Flex Western Starter Kit for fluorescent detection includes an iBind Flex Western Device , iBind Flex Cards, and an iBind Flex Fluorescent Detection (FD) Solution Kit.
Supplier: Invitrogen™ SLF2002S
Description
The iBind Flex Western Device is a small benchtop device that automates western blot processing (immunodetection). It applies mechanical pressure to an iBind Flex Card to generate sequential lateral flow (SLF) of immunodetection solutions for the blocking, antibody binding, and washing steps. The glass fiber matrix of the iBind Flex Card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.
The iBind Flex Western Device allows all solutions to be prepared and loaded in the device at the start of the procedure; all subsequent steps proceed automatically through SLF action—no electricity or batteries required. Blots are ready for the final detection steps in approximately 2.5 hours.
The iBind Flex Western Device includes three user-interchangeable well inserts to process up to one midi blot, two mini blots, or six vertically-cut strip blots when used with iBind Flex Cards, and the iBind Flex Solution Kit or iBind Flex Fluorescent Detection (FD) Solution Kit to process one mini-blot.
Specifications
| 1 iBind Flex Western Device, 10 iBind Flex Cards (store at room temperature), 1 iBind Flex Fluorescent Detection (FD) Solution Kit (store at 2 to 8°C) | |
| Fluorescence | |
| Nitrocellulose, PVDF | |
| ∼2.5 hr. | |
| Starter Kit | |
| Room Temperature |
| 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots | |
| Western Blot | |
| iBind | |
| 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots | |
| 1 kit |
For Research Use Only. Not for use in diagnostic procedures.