Learn More
IBI Scientific™ TUNAIR™ Shake Flask System
Catalog No. 15453315
Change view
Click to view available options
Product Type:
Dri-gauze; For 300mL flask
No. per Pack:
5
Catalog No. Product Type No. per Pack
15453315 Dri-gauze; For 300mL flask 5
1 options
Catalog No. 15453315 Supplier IBI Scientific™ Supplier No. SS3018
Only null left
Provides optimum growth conditions for aerobic microorganisms, mammalian cells and plant cells
  • Three flask design based on applications: No-Baffle (normal throw), Half-Baffle (vortex motion), and Full-Baffle (propeller motion)
  • Increased oxygen absorption rate due to unique baffling and turbo-vane closure design
  • Silicone or nitrocellulose filter linings are 0.22μ
  • Slip on two piece polypropylene cap eliminates need of flaming and protect neck of flask from airborne particles
  • Autoclavable
Applications:
Microbiology and biotechnology
Specifications
For Use With (Application) For 300mL Flask, Tunair shake flask system
No. per Pack 5
Product Type Dri-gauze; For 300mL flask