missing translation for 'onlineSavingsMsg'
Learn More

IBI Scientific™ Low Profile Roller Shaker Accessory, Replacement Roller

Catalog No. 361004440
Change view
Click to view available options
For Use With (Equipment):
IBI Low Profile Roller
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
361004440 IBI Low Profile Roller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361004440 Supplier IBI Scientific™ Supplier No. ACCRLSR10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with IBI Low Profile Roller

  • Roller dimensions: 1.12 in. dia. x 10 in.L (2.9 x 25.4cm)

Specifications

Product Type Replacement Roller
Material Rubber
Color Black
For Use With (Equipment) IBI Low Profile Roller