Learn More
IBI Scientific™ 3-Place Manifold for Culture Medium
For use with Flow Cells
Supplier:  IBI Scientific™ ACCFL0006
Catalog No. 15-453-235
Only null left
Explore available promotions
Includes: Three 6 in. (15.2cm) lengths of 1/8 in. (3.2mm) I.D. silicone tubing.
Description
Description
  • Allows delivery to three flow cells simultaneously
  • Consolidate effluent from multiple cells to waste bottle
Specifications
Specifications
3-Place Manifold for Culture Medium
Autoclavable
Flow Cells
Three 6 in. L of 0.12 in. I.D. Silicone tubing
Product Suggestions
Product Suggestions