Learn More
Therapak™ IATA Shipping Boxes for Ambient Transport
Catalog No. 22130050
Change view
Click to view available options
Dimensions (L x W x H) Exterior:
12.25 x 9.25 x 2.25 in.
5.5 x 4.125 x5.75 in.
6 x 4.25 x 4 in.
8 x 4.25 x 4 in.
Type:
Medium Ambient IATA Transport Box
Small Ambient IATA Transport Box
Tray Type IATA Shipping Box
Upright Small Box
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Exterior Type
22130050 6 x 4.25 x 4 in. Small Ambient IATA Transport Box
22130024 8 x 4.25 x 4 in. Medium Ambient IATA Transport Box
22130049 5.5 x 4.125 x5.75 in. Upright Small Box
22131487 12.25 x 9.25 x 2.25 in. Tray Type IATA Shipping Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22130050 Supplier Therapak™ Supplier No. 23675
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
  • Each box has a specific tab-lock mechanism to keep boxes closed during transport
  • Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
  • Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Compliance
Meets IATA Packing Instruction 650 dimensional requirements.
Specifications
Certifications/Compliance Meets IATA Packing Instruction 650 dimensional requirements.
Applications For shipment of Biological Substance Category B specimens
Size Small
Type Small Ambient IATA Transport Box
Dimensions (L x W x H) Exterior 6 x 4.25 x 4 in.
Dimensions (D x W x H) Interior 5.25 x 4 x 3.75 in.