Learn More
Therapak™ IATA Shipping Boxes for Ambient Transport
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
Supplier: Therapak™ TR23405
Description
- Each box has a specific tab-lock mechanism to keep boxes closed during transport
- Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
- Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Compliance
Meets IATA Packing Instruction 650 dimensional requirements.
Specifications
| Meets IATA Packing Instruction 650 dimensional requirements | |
| Small | |
| 5.5 x 4.125 x5.75 in. | |
| 200 test pound corrugated board |
| For diagnostic specimen shipments in IATA 650 compliance | |
| Upright Small Box | |
| 5.25 x 4 x 5.5 in. |