Learn More
Brady™ i7500 Printer Accessory Rewind Guide Plate
Description
- Replacement guide (Peel configuration printer ships with this guide as standard). Required when using peel configuration printer to internally rewind printed labels. Curved guide channels printed labels at a non-sharp angle back into printer for internal rewind. Guide has tabs that easily clip to front of the Peel configuration printer.
Specifications
Specifications
| Product Type | Guide Plate |
| For Use With (Equipment) | i7500 |