missing translation for 'onlineSavingsMsg'
Learn More

Cancer Diagnostics, Inc.™ i-Shields™ Eyeshields

Catalog No. 22500452
Encompass
Change view
Click to view available options
Color:
Assorted
No. per Pack:
LABPAK Professional Pack (25 Lenses and 10 Frames)
Product Type:
Eyeshield frame and lens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack Product Type
22500452 Assorted LABPAK Professional Pack (25 Lenses and 10 Frames) Eyeshield frame and lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22500452 Supplier Cancer Diagnostics, Inc.™ Supplier No. ESPP25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

10 Eyeshield FRAMES and 25 LENS.

Can be worn with a surgical mask.

  • Ultra Lightweight for stress free/mark free extended wearing
  • Reusable frame and disposable lenses provide side shield protection
  • Distortion-free, optical grade lenses work well with most sizes of prescription eyewear and can be worn with a surgical mask
  • Frames packaged in a variety of colors
  • Lightweight recyclable components can be easily sterilized

Order Info

NOTE: Lenses and frames are ordered separately unless ordered in combo pack (such as 22-500-452).

Specifications

Color Assorted
Product Type Eyeshield frame and lens
For Use With (Equipment) Most sizes of prescription eyewear and can be worn with a surgical mask
No. per Pack LABPAK Professional Pack (25 Lenses and 10 Frames)