A restriction endonuclease that recognizes the sequence TAGGG_ATAA^CAGGGTAAT. I-SceI is an intron-encoded endonuclease. The intron encoding I-SceI is present in the mitochondria of Saccharomyces cerevisiae.
Catalog No.
50812400
Supplier
New England Biolab LtdSupplier No.
R0694S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A restriction endonuclease that recognizes the sequence TAGGG_ATAA^CAGGGTAAT. I-SceI is an intron-encoded endonuclease. The intron encoding I-SceI is present in the mitochondria of Saccharomyces cerevisiae.