missing translation for 'onlineSavingsMsg'
Learn More

o-Phosphoric Acid, 85.0% W/W, MilliporeSigma™

Catalog No. 50948028
Change view
Click to view available options
Quantity:
4 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50948028 4 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50948028 Supplier Technidata Supplier No. PX09954000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH < 0.5 (100 g/L, H2O, 20°C)
Melting Point 21°C
Boiling Point 158°C (1013 mbar)
Packaging Plastic Bottle
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85.0% min
Quantity 4 L
Recommended Storage Store at room temperature