missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ miRNA Inhibitor, Negative Control #1

Catalog No. p-7075566
Encompass
Change view
Click to view available options
Quantity:
250 nmol
5 nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4464076 5 nmol
4464079 250 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4464076 Supplier Invitrogen™ Supplier No. 4464076
Only null left

mirVana miRNA Inhibitor Negative Control No. 1 is a non-targeting control for miRNA inhibition experiments. It provides a baseline for evaluating effects on gene expression. Extensively tested in human cells, it ensures no impact on known miRNA functions.

mirVana miRNA Inhibitor Negative Control No. 1 is a non-targeting control designed for use in miRNA inhibition experiments. It provides a reliable baseline for evaluating the effects on target gene expression by ensuring minimal off-target effects. Extensively tested in human cell lines and tissues, this control is validated to have no impact on known miRNA functions, making it essential for accurate and consistent gene expression studies. Its properties include high reliability, superior specificity, and versatile use in various experimental setups.

Features of mirVana miRNA Inhibitor Negative Control No. 1 include:

  • Reliability—maximum and consistent effect in vitro at low concentration and low toxicity
  • Potency—better potency than lead competitor's product
  • Superior specificity—the most updated Sanger content
  • Versatile use—can be used in vitro and in vivo, offering you consistency throughout your entire research project

What are mirVana miRNA Inhibitors?

mirVana miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity. Positive and negative controls are also available for use along with mirVana miRNA inhibitors in miRNA down-regulation experiments.

Quality control

Each mirVana miRNA Inhibitor is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each miRNA Inhibitor by gel electrophoresis to ensure a premium-quality miRNA inhibitor that is purified and ready for use.

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type miRNA Mimic
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line mirVana
Product Type miRNA Inhibitor
Purification Method Purified
Quantity 5 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.