Learn More
Pyramex™ I-Force™ Safety Eyewear
Catalog No. 296363033
Change view
Click to view available options
Lens Tint:
Amber
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
296363033 Clear
296363044 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296363033 Supplier Pyramex™ Supplier No. SB7010SDT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sporty, lightweight design eliminates the bulkiness and weight of a goggle
  • Ratcheting Quick Release interchangeable temples and strap
  • Vented foam carriage
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection.
  • H2X anti-fog lenses
  • Dual pane lens provides the ultimate anti-fog performance
  • Outer polycarbonate lens protects against the environment, while inner acetate lens is designed to prevent fogging
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Product Type Sealed Eyewear
    Frame Color Black
    Weight (Metric) 48 g
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate (Front Lens), Acetate (Inner Lens), PU Foam
    Frame Material Nylon
    UV Protection U6
    Frame Type Indirect Vented
    Show More Show Less