missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Burkholderia cepacia Selective agar plated medium (USP 60)

Catalog No. R110245
Change view
Click to view available options
85mm plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R110245 85mm plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R110245 Supplier Thermo Scientific™ Supplier No. R110245

Compliance to the USP 60 regulations for B. cepacia complex made simple

Burkholderia.cepacia Selective Agar Plated Medium (USP 60) is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in products, especially those for inhalation use of non-sterile aqueous preparations for oral, oromucosal, cutaneous or nasal use.

Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.

  • Compliant formulation according to USP 60
  • Quality tested to meet USP 60 specifically for Pharma customers
  • Part of a complete workflow which include DCM and QCO

Specifications

Description Burkholderia cepacia Selective (USP 60) Agar 10/PK
Format Standard petri dish
Product Type Agar Plate
For Use With (Application) Pharmaceutical testing of non-sterile products used for nasal inhalation
Quantity 10/Pk.
Regulatory Status Lab use only