missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyQ™-RS Reduced Serum Media

Catalog No. SH3056502
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Product Type:
Dulbecco's Modified Eagle Medium - Reduced Serum
Minimal Essential Medium - Reduced Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
SH3056502 1000 mL Dulbecco's Modified Eagle Medium - Reduced Serum
SH3056401 500 mL Minimal Essential Medium - Reduced Serum
SH3056501 500 mL Dulbecco's Modified Eagle Medium - Reduced Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3056502 Supplier Cytiva Supplier No. SH30565.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For applications requiring a lower concentration of serum

  • Support cell culture growth
  • Format: Liquid
  • Store at 2° to 8°C

Specifications

Classification None
Form Liquid
Product Type Dulbecco's Modified Eagle Medium - Reduced Serum
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
With Additives Glutamine, Glucose
Without Additives None
Quantity 1000 mL
Content And Storage 2° to 8°C
Description DMEM-RS with L-Glutamine