missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyQ™-RS Reduced Serum Media
For applications requiring a lower concentration of serum
Supplier: Cytiva SH30565.01
Description
- Support cell culture growth
- Format: Liquid
- Store at 2° to 8°C
Chemical Identifiers
| Liquid | |
| 0.1 μm Sterile-filtered | |
| 2° to 8°C |
| Reduced Serum Medium | |
| Glutamine, Glucose |
Specifications
| Liquid | |
| 0.1 μm Sterile-filtered | |
| 500 mL | |
| DMEM-RS with L-Glutamine |
| Reduced Serum Medium | |
| Glutamine, Glucose | |
| 2° to 8°C |
Safety and Handling
ShelfLife : 12 Months