missing translation for 'onlineSavingsMsg'
Learn More

Hypromellose, Substitution Type 2910, 4000 mPa.s, USP, 7-12%, Spectrum™ Chemical

Catalog No. 18603396
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603396 125 g Poly Bottle
18603397 500 g Poly Bottle
18603398 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603396 Supplier Spectrum Chemical Mfg Cor Supplier No. H1128125GM

H1128, 9004-65-3

Spectrum™ Chemical Hypromellose, Substitution Type 2910, 4000 mPa.s, USP is a semisynthetic, viscoelastic polymer that is inert and used as an ophthalmic lubricant, a controlled-delivery component and an excipient in oral medicaments. It is also used as a food additive and acts as an emulsifier, suspending agent and thickening agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
InChI Key PUSNGFYSTWMJSK-UHFFFAOYNA-N
IUPAC Name 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 7 to 12%
Grade USP
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Poly Bottle