missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine 98.0+%, TCI America™

Catalog No. p-7130481
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03115G 5 g
H031125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03115G Supplier TCI America Supplier No. H03115G

Chemical Identifiers

CAS 68-94-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005725
InChI Key FDGQSTZJBFJUBT-UHFFFAOYSA-N
Synonym hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
PubChem CID 790
ChEBI CHEBI:17368
IUPAC Name 6,7-dihydro-3H-purin-6-one
SMILES O=C1N=CNC2=C1NC=N2

Specifications

Quantity 5 g
Formula Weight 136.11
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Hypoxanthine
RTECSNumber UP0791000
TSCA Yes