Learn More
Stemcell Technologies Inc HypoThermosol FRS, Size: 500 mL
Catalog No. 501974436
Encompass
Improve and extend the preservation of cells, tissues, and organs with HypoThermosol FRS. This ready-to-use, serum-free, protein-free, and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 - 8C) preservation process. HypoThermosol FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility, endotoxin, and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers. Visit STEMCELL.com for more information.
Catalog No. 501974436 Supplier Stemcell Technologies Inc Supplier No. 07936
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Improve and extend the preservation of cells, tissues, and organs with HypoThermosol FRS. This ready-to-use, serum-free, protein-free, and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 - 8C) preservation process. HypoThermosol FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility, endotoxin, and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers. Visit STEMCELL.com for more information.