Learn More
Mopec™ Hypodermic Needles
Catalog No. 22445290
Change view
Click to view available options
Gauge:
12 G
13 G
15 G
19 G
20 G
Length (English):
1.5 in.
2.5 in.
3 in.
3.5 in.
4 in.
4.5 in.
5 in.
6 in.
8 in.
10 in.
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Length (English)
22445290 12 G 4 in.
22445354 12 G 5 in.
22445889 12 G 6 in.
22445651 13 G 4.5 in.
22445822 12 G 3 in.
22445785 13 G 8 in.
22445680 13 G 10 in.
22445552 15 G 3.5 in.
22445681 15 G 4 in.
22445783 15 G 5 in.
22445286 15 G 6 in.
22445531 19 G 3 in.
22445530 20 G 1.5 in.
22445551 20 G 2.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 22445290 Supplier Mopec™ Supplier No. FC91221
Hypodermic needles are designed for aspiration, cavity injection or extraction.
  • Long-lasting piercing points
  • Luer hubs
Specifications
Product Type Hypodermic Needle
Material Stainless Steel
Sterility Non-sterile
Gauge 12 G
Length (English) 4 in.