missing translation for 'onlineSavingsMsg'
Learn More

STERIS Hypochlorite WFI Sterile Solution

Catalog No. 19415126
Change view
Click to view available options
Volume (English):
22 oz.
32 oz.
1 gal.
Concentration:
0.525%
5.25%
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Concentration
19415126 1 gal. 0.525%
19415124 22 oz. 0.525%
19415125 32 oz. 0.525%
19415121 22 oz. 5.25%
19415122 32 oz. 5.25%
19415123 1 gal. 5.25%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19415126 Supplier STERIS Supplier No. 1S0808WR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxidizing cleaners can be extremely effective when removing tough soils and residue from hard surfaces, even the most critical surfaces

  • Designed to remove tough soil and residue from surfaces
  • Made with United States Pharmacopeia (USP) Water For Injection and Sodium Hypochlorite
  • Filtered, and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
  • Packages are double bagged for easy entry into controlled environments
  • Includes an extensive documentation package to meet validation objectives
  • Each lot documented by a Certificate of Analysis including sterility results
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
  • These products are not registered as disinfectants by the US EPA

Specifications

Product Type Bleach
Sterility Sterile
Composition Sodium Hypochlorite, USP Water For Injection (WFI)
Concentration 0.525%
Container Type Bottle
Filtered Filtered
For Use With (Application) Removing Tough Soil and Residue From Surfaces
Quantity 4/Cs.
Volume (English) 1 gal.