Learn More
Corning™ HYPERStack™ Manifold, MPC Connectors
GREENER_CHOICE
Catalog No. 15100190
Change view
Click to view available options
For Use With (Equipment):
(4) Corning™ HYPERStack™ cell culture vessels
Catalog No. For Use With (Equipment)
15100190 (4) Corning™ HYPERStack™ cell culture vessels
1 options
Catalog No. 15100190 Supplier Corning™ Supplier No. 11370
Only null left
For use with HYPERStack™ cell culture vessels.
Sterilization:
Irradiated and dosimetrically released based on ANSI/AAMI/ISO 11137 Sterilization of health care products- Requirements for validation and routine control-Radiation sterilization. Sterility Assurance Level (SAL) of 10-6.
Non-Pyrogenic:
Tested and met the criteria established in the current version of ANSI/AAMI ST 72, 'Bacterial Endotoxins - Test methodologies, routine monitoring, and alternative to batch testing'. The acceptance level for product is ≤ 0.1 EU/mL or ≤ 4 EU/device.
BSE/TSE:
Product complies with the latest revision of EMA/410/01 'Note for Guidance on minimizing the risk of transmitting animal spongiform encephalopathy agents via human and veterinary medicinal products' by virtue of all bovine derived material having been processed per specific conditions of section 6.4 of EMA/410/01.
Specifications
Material C-Flex™, Thermoplastic Elastomer
For Use With (Equipment) (4) Corning™ HYPERStack™ cell culture vessels
Includes PP clamp, C-Flex™ tubing, MPC connectors
Product Type HYPERStack™ Manifold