Learn More
Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns
Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.
Supplier:  Thermo Scientific™ 60311202
 View more versions of this product
Catalog No. 03150343
Only null left
Description
Description
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
Specifications
Chemical Identifiers
Conventional HPLC System
HyperSep
Online Solid Phase Extraction Cartridge
Pack of 4
Specifications
HyperSep Retain PEP; 3.0 mm ID
Conventional HPLC System
HyperSep Urea-Modified Polystyrene DVB
Pack of 4
Retention of Polar and Nonpolar Analytes
HyperSep
Online Solid Phase Extraction Cartridge
3.0 mm
Product Suggestions
Product Suggestions