Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates
Catalog No. 03251323
Change view
Click to view available options
Description:
Sample Collection Plate, 2 mL
Catalog No. Description
03251323 Sample Collection Plate, 2 mL
1 options
Catalog No. 03251323 Supplier Thermo Scientific™ Supplier No. 60300403
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates with your HyperSep system.
Use Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates with your HyperSep system.
Fast analysis without sacrificing accuracy or reproducibility. Incorporate traditional solid phase extraction sorbents into a 96-well plate format using the HyperSep system.
  • Innovative design of 96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation
  • Well plates are customizable to allow any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Ordering Information:
Available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Description Sample Collection Plate, 2 mL
For Use With (Application) SPE Sample Preparation
For Use With (Equipment) Vacuum Manifold
Product Line HyperSep
Type Sample Collection Plate
Volume (Metric) 2 mL