missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Retain PEP Plates

Catalog No. 60306207
Change view
Click to view available options
Bed Weight:
30 mg
60 mg
Volume (Metric):
1 mL
2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Volume (Metric)
60306207 30 mg 2 mL
60306208 60 mg 2 mL
03251495 30 mg 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60306207 Supplier Thermo Scientific™ Supplier No. 60306207

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get balanced retention for polar and nonpolar analytes with HyperSep Retain PEP Plates.

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System

Specifications

Description Retain PEP 30 mg/2 mL 96 Fixed Well Plate, 1 pk
Bed Weight 30 mg
For Use With (Application) Retention of Polar and Nonpolar Analytes
Particle Size 30 to 50 μm
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Solid Phase Extraction Well Plate
Volume (Metric) 2 mL
Volume (Metric) Well 1 mL
Quantity 1 Pack