missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate

Catalog No. p-7163716
Change view
Click to view available options
Bed Weight:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
60306503 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60306503 Supplier Thermo Scientific™ Supplier No. 60306503
Only null left

Unique material for retention of highly polar compounds.

Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents

  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules

Specifications

Bed Weight 50 mg
For Use With (Equipment) Universal Vacuum Manifold
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HypeSep Hypercarb
Type 96-well Plate
Quantity 1/Ea