Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Column Adapters
Catalog No. 03052531
Change view
Click to view available options
For Use With (Application):
SPE Sample Preparation
Catalog No. For Use With (Application)
03052531 SPE Sample Preparation
1 options
Catalog No. 03052531 Supplier Thermo Scientific™ Supplier No. 60104259
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Order these cartridge adapters when using various cartridge sizes on the glass block vacuum manifold.
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Cartridge Adaptors for using various sized solid phase extraction (SPE) cartridges on the glass block vacuum manifold for sample preparation.
Specifications
Description Column Adaptors for 1 mL, 3 mL , 6 mL SPE Columns, 15 pk
For Use With (Application) SPE Sample Preparation
For Use With (Equipment) Glass Block Vacuum Manifold, 1 mL, 3 mL , 6 mL SPE Columns
Product Line HyperSep
Type Column Adaptors
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.