Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Catalog No. 60105213
Change view
Click to view available options
Volume (Metric):
2 mL Centrifuge Tubes
6 mL Centrifuge Tubes
15 mL
For Use With (Application):
QuEChERS Method
Schenck Method
Description:
150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr
400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
750 mg anhydrous magnesium sulfate, 250 mg PSA, 250 mg C18, 250 mg GCB
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Application) Description
60105213 15 mL QuEChERS Method 750 mg anhydrous magnesium sulfate, 250 mg PSA, 250 mg C18, 250 mg GCB
03150629 6 mL Centrifuge Tubes Schenck Method 500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
03151608 6 mL Centrifuge Tubes Schenck Method 400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
60105232 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60105213 Supplier Thermo Scientific™ Supplier No. 60105213
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
Comprehensive range of Dispersive SPE products that contain the proper sorbents foroptimum extraction, clean-up and separation of analytes from complex matrices such as food using QuEChERS methods.
  • Save valuable time in preparation – products are supplied pre-prepared
  • Reduced variability due to consistency in products and rigorous quality control
  • Cleaner extracts from cleaner, pre-prepared products
  • Lower peak counts as a result of using pre-prepared products
Specifications
Description 750 mg anhydrous magnesium sulfate, 250 mg PSA, 250 mg C18, 250 mg GCB
For Use With (Application) QuEChERS Method
Product Line Quechers
Sorbent Anhydrous magnesium sulfate, PSA, C18, GCB
Type QuEChERS Sample Prep
Volume (Metric) 15 mL
Quantity 50
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.