missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep Carboxylic Acid (WCX) SPE Cartridge

Catalog No. 60108208
Change view
Click to view available options
Bed Weight:
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
10 mL
15 mL
Quantity:
100/pk
20/pk
30/pk
50/pk
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Column Capacity (Metric) Quantity
60108208 2000 mg 15 mL 20/pk
60108201 50 mg 1 mL 100/pk
60108202 100 mg 1 mL 100/pk
60108203 200 mg 3 mL 50/pk
60108207 200 mg 10 mL 50/pk
60108205 500 mg 6 mL 30/pk
60108206 1000 mg 6 mL 30/pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60108208 Supplier Thermo Scientific™ Supplier No. 60108208

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weak cation exchange sorbent for extraction of strong bases

SPE Tubes with frit . Hypersep Silica with WCX function for extraction of strong bases from biological fluids.

Specifications

Column Capacity (Metric) 15 mL
Bed Weight 2000 mg
Product Line HyperSep
Type SPE Cartridge
Quantity 20/pk