missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep Carboxylic Acid (WCX) SPE Cartridge

Catalog No. 60108201
Change view
Click to view available options
Bed Weight:
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
10 mL
15 mL
Quantity:
100/pk
20/pk
30/pk
50/pk
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Column Capacity (Metric) Quantity
60108201 50 mg 1 mL 100/pk
60108202 100 mg 1 mL 100/pk
60108203 200 mg 3 mL 50/pk
60108207 200 mg 10 mL 50/pk
60108205 500 mg 6 mL 30/pk
60108206 1000 mg 6 mL 30/pk
60108208 2000 mg 15 mL 20/pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60108201 Supplier Thermo Scientific™ Supplier No. 60108201

Weak cation exchange sorbent for extraction of strong bases

SPE Tubes with frit . Hypersep Silica with WCX function for extraction of strong bases from biological fluids.

Specifications

Column Capacity (Metric) 1 mL
Bed Weight 50 mg
Product Line HyperSep
Type SPE Cartridge
Quantity 100/pk