missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ C8 Cartridges

Catalog No. 03251295
Change view
Click to view available options
Bed Weight:
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
10 g
Column Capacity (Metric):
1 mL
3 mL
6 mL
15 mL
75 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Column Capacity (Metric)
03251295 10 g 75 mL
03251160 500 mg 3 mL
03251159 50 mg 1 mL
03251265 100 mg 1 mL
03251266 200 mg 3 mL
03251161 500 mg 6 mL
03251279 1000 mg 6 mL
03251293 2000 mg 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 03251295 Supplier Thermo Scientific™ Supplier No. 60108706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 cartridges offer excellent performance.

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ C8 Cartridges offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:

  • Drugs and their metabolites in biological samples
  • Peptides in biological samples

HyperSep C8 SPE Cartridges provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.

HyperSep SPE Cartridges and Plates

Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Specifications

Column Capacity (Metric) 75 mL
Description 10 g Bed Weight; 75 mL Column Volume
Bed Weight 10 g
For Use With (Application) Screening of Drugs, Metabolites, Peptides in Biological Samples
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep C8
Type Solid Phase Extraction Cartridge
Volume (Metric) 75 mL
Quantity Pack of 10
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.