missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Smartainer™ 3D BioProcess Container (BPC), 100L
GREENER_CHOICE

Catalog No. SH3071701
Change view
Click to view available options
Quantity:
100 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3071701 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3071701 Supplier Thermo Scientific™ Supplier No. SH30717.01

The 100 L Thermo Scientific large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports (2 MPX inserts), 1 bottom drain (MPX insert).

The 100 L Thermo Scientific™ large-volume BPC, 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports (2 MPX inserts), 1 bottom drain (MPX insert).

Features:

  • Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
  • All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
  • All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:

  • Hydration and sterilization of process buffers, liquids, and culture media.
  • Chromatography feed and fraction collection.
  • Storage and transport of bulk drug product and bulk drug precursors.
  • Harvest from and feed into bioreactors and fermentors.
  • Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology

Choose from the industry's largest component library – with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.

Specifications

Capacity (Metric) 100 L
Film Description CX5-14
Port Detail Bottom Port 1 x 0.5 in., Top Ports: 2 x 0.5 in.
For Use With (Equipment) Smartainer™ Hardware Stainless Steel Outer Container (Cat. No. SV50160.01)
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 100 L
Sterilization Method Gamma Irradiated
Tubing Length Line 1 51 cm (20 in.)
Tubing Length Line 2 51 cm (20 in.)
Tubing Length Line 3 51 cm (20 in.)
Tubing Line 1 MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Top Port
Tubing Line 2 MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Top Port
Tubing Line 3 MPX Body, Polycarbonate, ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) Bottom Port
Tubing Material Line 2 Silicone
Tubing Material Line 1 Silicone
Tubing Material Line 3 Silicone
Product Line HyPerforma™, Smartainer™ II
Show More Show Less