Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Catalog No. sh3118801
Change view
Click to view available options
Capacity (Metric):
200 L
500 L
Insert Type:
Horizontally-gusseted
Vertically-gusseted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Insert Type
SH3118801 200 L Horizontally-gusseted
SH3118802 500 L Horizontally-gusseted
SH3118803 500 L Vertically-gusseted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3118801 Supplier Thermo Scientific™ Supplier No. SH31188.01
Only null left
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
Specifications
Capacity (Metric) 200 L
Product Type Bioprocess Container (BPC)
Insert Type Horizontally-gusseted