Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
$1,969.34 - $2,131.16
| Catalog Number | Mfr. No. | Capacity (Metric) | Insert Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Insert Type | Price | Quantity & Availability | ||||
|
SH3118801
|
Thermo Scientific™
SH31188.01 |
200 L | Horizontally-gusseted |
Each for $1,969.34
|
|
||||
|
SH3118802
|
Thermo Scientific™
SH31188.02 |
500 L | Horizontally-gusseted |
Each for $2,131.16
|
|
||||
|
SH3118803
|
Thermo Scientific™
SH31188.03 |
500 L | Vertically-gusseted |
Each for $1,989.20
|
|
||||
Description
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.Specifications
| Bioprocess Container (BPC) |