missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Mixtainer™ Hardware, Conical Drum, 200L
GREENER_CHOICE

Catalog No. SV5051713
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV5051713 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SV5051713 Supplier Thermo Scientific™ Supplier No. SV5051713

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The 200L Thermo Scientific™ Conical Drum top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications.

The 200L Thermo Scientific™ Conical Drum top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a magnetic stirring plate, a support plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.

Recommended for:

  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions

Specifications

Capacity (Metric) 200 L
Product Line HyPerforma, Mixtainer
Port Detail Bottom Port: 4 in. (10.2 cm)
Quantity 200 L
Dimensions 60 cm x 114 cm (23.5 in. x 44.75 in.)