missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Conical LLDPE Drum, 50L, 1 port, with clamps

Thermo Scientific™ HyPerforma Conical LLDPE Drums are designed for use with BioProcess Container Systems and Tank Liners.

Supplier:  Thermo Scientific™ SV5051708

Catalog No. SV5051708


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ HyPerforma™ Conical LLDPE Drums are designed for use with BioProcess Container™ Systems and Tank Liners.

  • Includes lid and clamps
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications

Specifications

Conical LLDPE 4
LLDPE
60 x 58 cm (23.5 x 23 in.)
50 L
Fewer resources used and less waste
50 L
Bottom Port: 10.2 cm (4 in.)
HyPerforma
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions