missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Conical LLDPE Drum, 200L, 1 port, with clamps
GREENER_CHOICE

Catalog No. SV5051710
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SV5051710 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SV5051710 Supplier Thermo Scientific™ Supplier No. SV50517.10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ HyPerforma Conical LLDPE Drums are designed for use with BioProcess Container Systems and Tank Liners.

Thermo Scientific™ HyPerforma™ Conical LLDPE Drums are designed for use with BioProcess Container™ Systems and Tank Liners.

  • Includes lid and clamps
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port

Specifications

Description Conical LLDPE 4
Material LLDPE
Capacity (Metric) 200 L
Dimensions 60cm x 114cm (23.5 in. x 44.75 in.)
Port Detail Bottom Port: 10.2 cm (4 in.)
Quantity Each
Product Line HyPerforma