missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 100L
GREENER_CHOICE

Catalog No. SH3064702
Catalog No. SH3064702 Supplier Thermo Scientific™ Supplier No. SH30647.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Tank Liner Key Features:

  • Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
  • Gusset design allows for a close fit to the tank
  • Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
  • Supplied gamma irradiated
  • Customization options are available by contacting one of our Thermo Scientific Representatives today

Standard Tank Liners for your liquid applications available in-stock in the following configurations:

SH30647.01 to .03:

  • 50 to 200L Tank Liner
  • Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
  • Quantity: Each = 1 Liner

SH30647.04 to .07:

  • 50 to 500L Tank Liner
  • Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
  • Quantity: Each = 1 Liner

SH30646 Series:

  • 50 to 200L Tank Liner with One Bottom Drain
  • Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
  • Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)

Ordering Information:

Supplied double polybagged and irradiated with Certificate of Analysis.

Recommended for:

  • Hydrations of powdered media and buffers
  • Pooling of non-sterile solutions and fluids
  • Non-aseptic collection of waste liquids from separation and purification systems

To order, use the part numbers listed in the above table.

Specifications

Capacity (Metric) 100 L
Material Plastic
For Use With (Application) Handling Fluids in Bioproduction Application
Product Type Tank Liner
Outer Packaging Packages are placed in strong cardboard cartons with product and lot identification.
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
For Use With (Equipment) SV50076.03
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9
Sterility Sterile
Show More Show Less