missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood
Protect users and the environment from hazardous vapors generated on the work surface with our series of high-efficiency Epredia™ Hyperclean™ TruAir™ Ductless Fume Hoods.
Supplier: Epredia™ B1115483
Description
Innovation in air purification, unmatched air quality- Protects the operator from fume and particle hazards
- Easy to change filters
- Improved filter clamping eliminates by-pass leakage
- Low airflow alarm
Advanced carbon filtration technology offers a safe, high performance alternative to conventional ducted fume hoods for a broad range of applications.
- Environmental Benefits: Epredia ductless fume hoods isolate and trap chemical vapors to prevent ecological impact through release into the environment
- Versatile: Each filtration system is selected for its specific application. Carbon filters are available in several configurations for use with vapors or organic solvents, acids, mercury and formaldehyde. HEPA filters can add to biological safety
- Easy to Install: The ductless fume hood is self-contained and does not require venting to the outside. Many units are portable and may be moved from one location to the next with minimal down-time and without filter changes. Set up, operation and filter maintenance are straightforward
- Energy Efficient: Because filtered air is returned to the room, no demands are required of the facility HVAC capacity for make-up air
- Cost Effective: Facility ductwork, HVAC and construction costs are eliminated
- Safe to Use: Cabinet airflow and face velocity protect users from incidental exposures to fumes
- Self testing: Electronic airflow monitoring assures continuous safety
Product Features
- Filter I.D. Window: A strategically placed front cover window shows the installed filter part number and installation date for convenience and to encourage timely filter replacement.
- Hinged Front Sash: When closed, the cabinet sash protects the contents from inadvertent external contact, and better isolates the air within. The sash is easy to open and close.
- Control Panel: Electronic controls and displays include switches for the blower and low airflow alarm.
- Steel Support Frame: The chemical resistant epoxy coated steel frame adds mechanical strength. The pre-filter can be changed while the unit is operating to prevent operator exposure to chemical vapors.
- Electrostatic Pre-Filter: The 99.5% effective electrostatic pre-filter is accessible from inside the chamber to contain the release of any particulates that it traps.
- Pass Through Ports: Electrical cords and cables are safely routed into the cabinet through ports on the back and side walls.
- Color: The cabinet is light gray; side and back panels are clear.
- Airflow Alarm: A continuous air velocity monitoring system alerts the operator upon unacceptable values.
- Internal Manual Speed Controller: Authorized personnel may set the centrifugal fan motor speed as desired.
- Stand: Optional mobile cart with locking casters.
- Work Surface: The internal work surface can be fitted with an optional polypropylene tray (see Accessories).
- Filter Door Key: Filter access keys prevent unauthorized removal or accidental exposure to dirty filters.
Chemical Identifiers
| Ductless Fume Hood | |
| Steel | |
| 2 Years |
| Compact Fluorescent | |
| Hyperclean™, TruAir™ |
Specifications
| Ductless Fume Hood | |
| Steel | |
| 67.6 cm | |
| 281.25 cfm | |
| 35 in. | |
| 78.1 cm | |
| 120 V | |
| 138 lb. | |
| 48 in. |
| Compact Fluorescent | |
| 27 in. | |
| Extra Tall Standard Leg 48 | |
| Hyperclean™, TruAir™ | |
| 24 in. | |
| 61 cm | |
| 2 Years | |
| 63 kg | |
| 121.9 cm |
Some assembly may be required.