Learn More
Brands HYP MOS 5 4X2MM GRD CRT 3
Catalog No. 50926703
Encompass
Agilent LS Hyp MOS, 5u, 4.0 x 20mm guard cart, 3/pk Hypersil-BC
Catalog No. 50926703 Supplier Brands Supplier No. 79916KT121
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Agilent LS Hyp MOS, 5u, 4.0 x 20mm guard cart, 3/pk Hypersil-BC