missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B

Catalog No. AAJ60681MD
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60681MD 250 mg
AAJ60681MC 100 mg
AAJ6068103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60681MD Supplier Thermo Scientific Chemicals Supplier No. J60681MD

Hygromycin B, CAS # 31282-04-9, is an aminoglycoside compound showing antibiotic activity that kills bacteria through the inhibition of protein synthesis. It also presents anthelmintic activity. | CAS: 31282-04-9 | C20H37N3O13 | 527.524 g/mol

An aminoglycoside antibiotic that kills bacteria by inhibiting protein synthesisHygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contains the hygromycin resistance gene. This gene is frequently used as a selectable marker in research on plants. It stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. In addition to this, it serves as an aminoglycosidic antibiotic which inhibits protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hygromycin is an aminoglycosidic compound that shows antibiotic activity. It is produced by Streptomyces hygroscopicus
  • Hygromycin acts against both prokaryotic and eukaryotic cells, having the ability to inhibit protein synthesis. It also stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. This compound prevents ribosome translocation during translation elongation by binding the 30S rRNA proximal to the ribosomal E, P, and A sites

Application

  • In clinical and plant research, this compound can be used as a marker to select and maintain prokaryotic and eukaryotic cells containing the hygromycin resistance gene
  • Hygromycin also presents an anthelmintic activity against nematode parasites of chickens

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
MDL Number MFCD06795479
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Lyophilized Powder
Color Brown
Quantity 250 mg
UN Number UN3462
Merck Index 14,4852
Solubility Information Miscible with water,methanol,buffer solution and ethanol; Immiscible with less polar solvents.
Formula Weight 527.53
Chemical Name or Material Hygromycin B
Hazard Category H300+H310+H330-H317-H318-H334
Hazard Statement GHS H Statement
H300-H310-H330-H334-H318-H317
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P262-P264b-P270-P271-P272-P280-P284-P285-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
EINECSNumber 250-545-5
RTECSNumber WK2130000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only