missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B Solution, ≥80%, MP Biomedicals™

Catalog No. ICN15751394
Change view
Click to view available options
Quantity:
5 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15751394 5 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15751394 Supplier MP Biomedicals Inc Supplier No. 0215751394

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An aminoglycoside antibiotic produced by the bacterium Streptomyces hygroscopicus that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
MDL Number MFCD06795479
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
Synonym hygromycin b
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Color Yellow
Assay Percent Range ≥80%
Quantity 5 mU
Formula Weight 527.5
Physical Form Liquid
Chemical Name or Material Hygromycin B Solution