Learn More
Hygiena SystemSURE Plus™ ATP Monitoring System
Catalog No. 23230100
Change view
Click to view available options
Type:
ATP Monitoring system
Catalog No. Type
23230100 ATP Monitoring system
1 options
Catalog No. 23230100 Supplier Hygiena Supplier No. SS3H
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Carrying case with a shoulder strap for hands-free transportation to test locations in your facility.
Easy-to-use, handheld device uses state-of-the-art technology to perform ATP testing to determine the hygienic status of surfaces and liquids.
  • Extremely sensitive to low levels of contamination
  • Advanced photodiode technology - Internal solid state detector is not affected by drops or shakes
  • Storage: 100 programmable test plans; 200 programmable user IDs; 251 programmable locations per test plan; 5000 programmable test locations
  • Small, lightweight, handheld instrument
  • Quick-test mode enables ad-hoc testing
  • Large screen with back-light and optional BOLD text
  • Powered by two AA batteries for several months of uninterrupted use
  • Connects to software via USB
  • Removable read chamber design allows for easy cleaning
  • Durable outside shell and sealed keypad to protect against splashes and wear
Advantages
  • Cost-effective ATP hygiene monitoring system
  • Provides real-time feedback and facilitates long-term data analysis
  • Optimizes and verifies that cleaning procedures are working
  • No scheduled yearly maintenance necessary
  • Available Calibration Control Kit for in-house confirmation of calibration
  • Compatible with various tests for surfaces, water samples, and endoscope sampling
  • Complimentary SureTrend Software
Warranty
One year; includes parts and labor.
Specifications
Type ATP Monitoring system
Connection Type USB
Range 1 femtomole (1 x 10-15 moles) of ATP
For Use With (Application) Evaluate effectiveness of disinfectants and sanitizer, measure performance of housekeeping staff and to ensure compliance with cleaning protocols
Height (Metric) 17.8 cm
Height (English) 7 in.
Width (English) 3 in.
Width (Metric) 7.6 cm
Includes Carrying case with a shoulder strap