missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-400 Medium Duty Nitrile Coated Oil Repellent Gloves

Catalog No. 19019860
Change view
Click to view available options
Size:
8
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19019860 8
19019865 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19019860 Supplier Ansell Supplier No. 4740011080
Only null left

ActivArmr™ gloves combine the comfort of cotton or light leather gloves with the abrasion resistance and oil repellence of nitrile coated gloves.

  • ActivArmr™ gloves feature interlock knit lining, generous sizing, and contoured designs for improved comfort
  • Special nitrile formulation provides a combination of flexibility, mechanical resistance, and oil repellency
  • Natural hand shape is easier to wear for long periods
  • Gloves ensure cleanliness of metal sheets in the steel and motor industries
  • Silicone-free

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 47-400
Show More Show Less