missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11842 Nylon Multi-Purpose Gloves

Catalog No. 18000051
Change view
Click to view available options
Size:
10
11
12
5
6
7
8
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18000051 11
18000054 5
18000055 6
18000056 7
18000057 8
18000050 9
18000053 10
18000052 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18000051 Supplier Ansell Supplier No. 11842110
Only null left

HyFlex™ 11842 Nylon Multi-Purpose Gloves are sustainable gloves made with recycled nylon yarn.

  • Sustainable design: HyFlex™ 11-842 sustainable Multi-Purpose glove liners are designed with 90% recycled nylon yarn* reducing our carbon footprint by 75g CO2eq/pair** and plastic-free inner packaging that contains 53% fewer materials.^
  • Increased durability: FORTIX™ Technology provides up to 20% greater abrasion resistance.^^
  • Enhanced comfort: ZONZ™ Comfort Fit Technology helps improve breathability and dexterity.
  • Touchscreen compatibility: These industrial gloves work with multiple touchscreen devices - no need to remove them when switching between tasks for enhanced workplace safety
  • Certified skin-friendliness: These HyFlex™ gloves are dermatologically tested and free of harmful substances.

*minimum recycled content of more than 30% in the final product (based on size 9)

**vs. standard nylon multi-purpose gloves

^71% less greenhouse gas emissions - Ansell's LCA, calculated as weight reduction, peer reviewed

^^vs. previous technology version

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance Ecovadis label, EN ISO 21420:2020, EN 388:2016, EN 388:2016 +A1:2018, EN407:2020, ANSI ABR 4, Food Contact, CE Number, Dermatologically Tested, OEKO-TEX™ Centexbel
Coated Area Palm
Coating Material Foam Nitrile
Color Green
Cut Resistance Rating ANSI A4
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material Recycled Nylon
Product Type Foam Nitrile Gloves
Size 11
Glove Cuff Style Knit
Show More Show Less